Would you like to log in again?
HiScreen Capto DEAE Columns
Supplier: Cytiva
HiScreen™
28926982 28-9269-82
97067-804EA 197.75 USD InStock
97067-804 CA97067-804
HiScreen Capto DEAE Columns
Chromatography Columns
HiScreen™ Capto™ DEAE column is a weak anion exchanger with a bed height of 10 cm making it an excellent tool for method optimization, parameter screening, robustness testing, and convenient scale-up.
  • Prepacked, ready-to-use columns for convenient process development optimization
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Easily connected in series to achieve 20 cm bed height
  • Reproducible results, scalable to BioProcess columns packed with the same media using the same linear fluid velocity
  • Used in an optimal way with liquid chromatography systems such as ÄKTA™
  • Flow velocity: <700 cm/h
Process fluid velocities can be applied, since the bed height gives enough residence time and the results can then serve as basis for linear process scale-up. It is prepacked with Capto™ DEAE ion exchange chromatography medium.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty