Would you like to log in again?
Heparin Sepharose™ 6 Fast Flow Affinity Chromatography Media, Cytiva
Supplier: Cytiva
Sepharose™
17-0998-01 17-0998-01 17-0998-01
95017-066EA 1048.6 USD InStock
95017-066 97067-842 CA95017-066
Heparin Sepharose™ 6 Fast Flow Affinity Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Heparin Sepharose 6 Fast Flow is composed of cross-linked 6% agarose beads modified sodium heparin. Immobilization is performed using chemically stable and oriented coupling giving high binding capacity. The chemical stability of the media is solely dictated by the heparin ligand. Heparin Sepharose 6 Fast Flow is a member of the Cytiva range of affinity chromatography media for capture and intermediate purification.
  • For affinity chromatography
  • Industrial purification of antithrombin III, coagulation factors and other plasma proteins
  • Coupling chemistry gives chemically-stable ligand attachment
  • Reductive amination coupling method
  • Stability of medium solely dictated by heparin ligand
  • BioProcess medium supported for industrial applications and well-established in approved processes
  • Stable in commonly used aqueous buffers 0.1 M NaOH, 0.05 M sodium acetate, 8 M urea, 6 M guanidine hydrochloride, non-ionic detergents, 10% glycerol
Heparin Sepharose 6 Fast Flow is composed of cross-linked 6% agarose beads modified sodium heparin
This medium allows for fast and reliable separations of biomolecules with an affinity for heparin, including antithrombin III, coagulation factors and other plasma proteins. Additional applications include DNA binding proteins, lipoproteins, protein synthesis factors, enzymes that act on nucleic acids, and steroid receptors.
Heparin Sepharose 6 Fast Flow is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, Heparin Sepharose 6 Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.
Immobilization is performed using chemically stable and oriented coupling giving high binding capacity. The chemical stability of the media is solely dictated by the heparin ligand. Heparin Sepharose 6 Fast Flow is a member of the Cytiva range of affinity chromatography media for capture and intermediate purification.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty