Would you like to log in again?
Phenyl Sepharose™ High Performance Hydrophobic Interaction Chromatography Media, Cytiva
Supplier: Cytiva
Sepharose™
17-1082-01 17108201
97067-872EA 691.47 USD InStock
CA97067-872 97067-872
Phenyl Sepharose™ High Performance Hydrophobic Interaction Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Phenyl Sepharose High Performance is an aromatic hydrophobic interaction chromatography (HIC) medium, designed for intermediate and polishing step purification steps when high resolution has priority.
  • Aromatic HIC medium designed for intermediate purification and polishing steps
  • Selectivity similar to Phenyl Sepharose 6 Fast Flow (low sub) but higher resolution due to the smaller 34 µm bead size
  • BioProcess medium supported for industrial applications and well-established in approved processes
  • The hydrophilic nature of the base matrix ensures low levels of non-specific binding
  • Stable in commonly used aqueous buffers
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty