Would you like to log in again?
Capto™ S Ion Exchange Chromatography Media
Supplier: Cytiva
Capto™
17544110 17-5441-10 17544101 17-5441-01
97067-888EA 401.05 USD InStock
97067-888 CA97067-888 CA97067-890 97067-890
Capto™ S Ion Exchange Chromatography Media
Protein Purification Protein Purification Resins
Capto™ S is a strong cation exchange bioprocessing medium, designed to meet the demands of modern large-scale manufacturers for fast, efficient, and cost-effective capture and intermediate protein purification when high volume throughput is essential.
  • Strong cation exchanger for high-productivity capture and intermediate purification when high volume throughput is essential
  • High dynamic binding capacity at high flow raises productivity
  • High-volume throughput cuts process times
  • Cost-effective processing with smaller unit operations
Capto S withstands effective and rigorous CIP procedure and the chemical stability ensures long media lifetime. Capto S is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, Capto S meets industrial demands with security of supply and comprehensive technical and regulatory support.
Capto S is designed for capture and intermediate purification of proteins from large feed volumes. It is composed of a rigid, high-flow agarose matrix modified with dextran surface extenders and a strong sulfonate (S) cation exchanger. The combination of high volume throughput and high dynamic binding capacity for Capto S raises the productivity in industrial downstream processes. Capto S withstands effective and rigorous CIP procedure and the chemical stability ensures long media lifetime.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty