Would you like to log in again?
Capto™ DEAE Ion Exchange Chromatography Media, Cytiva
Supplier: Cytiva
Capto™
17544301 17-5443-01 17-5443-10 17544310
97067-894EA 205.78 USD InStock
CA97067-894 CA97067-892 97067-894 97067-892
Capto™ DEAE Ion Exchange Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Capto™ DEAE is a strong anion exchange bioprocessing medium, designed to meet the demands of modern large-scale manufacturers for fast, efficient, and cost-effective capture and intermediate protein purification when high volume throughput is essential.
  • High dynamic binding capacity at high flow raises productivity
  • High volume throughput cuts process times
It withstands effective and rigorous CIP procedure and the chemical stability ensures long media lifetime.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty