Would you like to log in again?
MabSelect Xtra™ Affinity Chromatography Media, Cytiva
Supplier: Cytiva
MabSelect™
17-5269-02 17526902 17-5269-07 17526907
97067-910EA 3887.04 USD InStock
CA97067-908 97067-910 CA97067-910 97067-908
MabSelect Xtra™ Affinity Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
MabSelect Xtra has the same recombinant Protein A ligand as MabSelect but is based on a high-flow agarose base matrix with increased porosity and a slightly decreased particle size compared to MabSelect.
  • Higher dynamic binding capacity than currently available Protein A chromatography media
  • High purity capture due to high ligand selectivity and matrix hydrophilicity
  • Epoxy activation coupling method
  • Slightly lower pressure/flow properties compared to MabSelect SuRe and MabSelect
  • Improved process economics through reduced raw materials costs and/or reduced number of cycles
  • Prepacked, ready-to-use columns for convenient process development
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
MabSelect Xtra is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, MabSelect Xtra meets industrial demands with security of supply and comprehensive technical and regulatory support. The medium provides a lower overall production cost due to the possibility of processing concentrated feedstocks in fewer batches.
This results in higher capacity—MabSelect Xtra has about 30% higher dynamic binding capacity for IgG than currently available Protein A chromatography media. In addition, MabSelect Xtra has a higher capacitiy for many Fc-fusion proteins.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty