Would you like to log in again?
Capto™ Phenyl (High Sub) Hydrophobic Interaction Chromatography Media, Cytiva
Supplier: Cytiva
Capto™
17-5451-01 17545101
97067-924EA 274.33 USD InStock
CA97067-924 97067-924
Capto™ Phenyl (High Sub) Hydrophobic Interaction Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Capto™ Phenyl (high sub) is used in the capture and intermediate stages of protein purification
  • Improved productivity and process economy in downstream operations
  • Very high flow rates and large sample volume processing
  • Excellent chemical stability
The combination of high capacity, narrow specification range, high flow rate and low backpressure consistently reduces process cycle times and increases productivity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty