Would you like to log in again?
Capto™ Q ImpRes Ion Exchange Chromatography Media, Cytiva
Supplier: Cytiva
Capto™
17-5470-02 17547010 17547002 17-5470-10
97067-936EA 579.67 USD InStock
97067-936 CA97067-938 97067-938 CA97067-936
Capto™ Q ImpRes Ion Exchange Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Capto Q ImpRes is a strong anion exchange BioProcess medium, designed to meet the demands of modern large-scale manufacturers for fast, efficient, and cost-effective intermediate and polishing protein purification.
  • High-resolution intermediate purification and polishing based on the well-established Capto platform with traditional ligands
  • Flexible process design due to large operational window of flow rates and bed heights
  • High-throughput purifications easy to optimize and scale up
  • Higher manufacturing productivity enables improved process economy
  • Security of supply and comprehensive regulatory support
Capto Q ImpRes is a strong anion exchanger for the high-throughput intermediate purification and polishing steps of a wide range of biomolecules. This chromatography media is part of an expanded high-resolution platform based on the high-flow agarose Capto product line.
By combining the high-flow characteristics of Capto media with a small particle size, Capto Q ImpRes delivers excellent pressure-flow properties with impressive resolution. The ability to run at higher flow rates and higher bed heights also increases flexibility in process design.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty