Would you like to log in again?
DEAE Sepharose™ Fast Flow Ion Exchange Chromatography Media, Cytiva
Supplier: Cytiva
Sepharose™
17-0709-01 17070910 17-0709-10 17-0709-01
97067-950EA 884.76 USD InStock
97067-950 97067-972 CA97067-972 CA97067-950
DEAE Sepharose™ Fast Flow Ion Exchange Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
DEAE Sepharose Fast Flow is a weak anion exchanger based on the well established Sepharose Fast Flow ion exchange platform, extensively used for preparative protein separations in both research and industrial applications.
  • Well-proven weak anion exchanger developed for industrial downstream processes
  • Used extensively for capture and intermediate purification of a wide range of approved biopharmaceuticals
  • The industry standard for ion exchange chromatography during recent decades
  • High chemical stability allows for well proven CIP and sanitization protocols
  • The hydrophilic nature of the base matrix ensures low levels of non-specific binding leading to low levels of host cell-derived impurities in the elution pool
  • Stable to all commonly used aqueous buffers additives such as 8 M urea and 6 M guanidine hydrochloride
DEAE Sepharose Fast Flow has high chemical stability, allowing well proven cleaning-in-place (CIP) and sanitization protocols. Scale-up with DEAE Sepharose Fast Flow is straight forward and the medium is available in a range of pre-packed process development tools. As member of the BioProcess media range, DEAE Sepharose Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.
DEAE Sepharose Fast Flow is part of the Sepharose Fast Flow ion exchange platform, which has been the industrial standard for ion exchange chromatography during recent decades. It is composed of crosslinked 6% agarose beads, with diethylaminoethyl (DEAE) weak anion exchange groups.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty