Would you like to log in again?
Phenyl Sepharose™ 6 Fast Flow (Low Sub) Hydrophobic Interaction Chromatography Media, Cytiva
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sepharose™
17-0965-10 17-0965-05 17096505 17096510
97067-978EA 224.13 USD InStock
CA97067-956 CA97067-978 97067-978 97067-956
Phenyl Sepharose™ 6 Fast Flow (Low Sub) Hydrophobic Interaction Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Phenyl Sepharose 6 Fast Flow is a well established, standard aromatic hydrophobic interaction chromatography (HIC) resin for capture and intermediate purification requiring low to resin hydrophobicity
  • Standard aromatic HIC resins based on Sepharose Fast Flow base matrix derivatized via uncharged, chemically-stable ether linkages.
  • Designed for initial and intermediate purification requiring low to medium hydrophobicity
  • Two levels of substitution help find optimal selectivity and binding capacity for a given application
  • BioProcess resin supported for industrial applications and well-established in approved processes
  • The hydrophilic nature of the base matrix ensures low levels of non-specific binding.
Phenyl Sepharose 6 Fast Flow is part of the Sepharose Fast Flow HIC platform, which has been an industrial standard for HIC chromatography during recent decades. Phenyl Sepharose 6 Fast Flow is composed of cross-linked, 6% agarose beads modified with standard aromatic phenyl groups via uncharged, chemically-stable ether linkages. Two levels of ligand substitution degree help to find the optimal selectivity and binding capacity for a given application.
Phenyl Sepharose 6 Fast Flow is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess resins range, Phenyl Sepharose 6 Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty