Would you like to log in again?
Butyl-S Sepharose™ 6 Fast Flow Hydrophobic Interaction Chromatography Media, Cytiva
Supplier: Cytiva
Sepharose™
17097802 17-0978-02
97067-958EA 1121.24 USD InStock
97067-958 CA97067-958
Butyl-S Sepharose™ 6 Fast Flow Hydrophobic Interaction Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Butyl-S Sepharose™ 6 Fast Flow is composed of cross-linked 6% agarose beads modified with butyl groups via a thio-ether linkage. This resin is designed for the binding and elution of relatively strong hydrophobic molecules at comparatively low salt concentrations.
  • Binds and elutes relatively strong hydrophobic molecules at comparatively low salt concentrations
  • The least hydrophobic medium in the well proven Sepharose Fast Flow HIC range
  • Purifies recombinant Hepatitis B virus surface antigen from CHO cells
  • Developed and optimized in cooperation with a process-scale manufacturer of biopharmaceuticals
  • The hydrophilic nature of the base matrix ensures low levels of non-specific binding
  • The least hydrophobic medium in the well proven Sepharose Fast Flow HIC range
This resin is designed for the binding and elution of relatively strong hydrophobic molecules at comparatively low salt concentrations. Butyl‑S Sepharose 6 Fast Flow is particularly useful during the initial stages of a separation process to remove the bulk of impurities without stringent requirements for conditioning of the sample (e.g. adjustment of pH, salt concentration and so on). Butyl-S Sepharose 6 Fast Flow is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, Butyl-S Sepharose 6 Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.
Butyl-S Sepharose 6 Fast Flow is part of the Sepharose Fast Flow HIC platform, which has been an industrial standard for HIC chromatography during recent decades. Butyl-S Sepharose 6 Fast Flow is composed of cross-linked 6% agarose beads modified with butyl groups via a thio-ether linkage. It will exhibit a different selectivity compared to Butyl Sepharose 4 Fast Flow and other HIC Sepharose Fast Flow products.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty