Would you like to log in again?
Sepharose™ 4 FF Octyl Fast Flow Hydrophobic Interaction Chromatography Media, Cytiva
Supplier: Cytiva
Sepharose™
17094602 17-0946-02
97067-964EA 1012.62 USD InStock
CA97067-964 97067-964
Sepharose™ 4 FF Octyl Fast Flow Hydrophobic Interaction Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Octyl Sepharose™ 4 Fast Flow resin is based on cross-linked 4% agarose. The aliphatic ligand is immobilised to the base matrix with an ether linkage. The ligand contains no charged groups, making true hydrophobic interaction chromatography possible, without interfering ionic effects.
  • Aliphatic HIC media based on Sepharose Fast Flow base matrix derivatised via uncharged, chemically-stable ether linkages
  • Optimised for the separation of larger proteins in capture and intermediate purification steps
  • Bioprocess medium supported for industrial applications and well established in approved processes
This resin is based on cross-linked 4% agarose. The aliphatic ligand is immobilized to the base matrix with an ether linkage. The ligand contains no charged groups, making true hydrophobic interaction chromatography possible, without interfering ionic effects. Octyl Sepharose 4 Fast Flow is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, Octyl Sepharose 4 Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.
The octyl derivative adds a different and complementary selectivity to the Sepharose Fast Flow HIC product family. Optimized for the separation of larger proteins in capture and intermediate purification steps. BioProcess medium supported for industrial applications and well established in approved processes. Octyl Sepharose 4 Fast Flow is part of the Sepharose Fast Flow HIC platform, which has been an industrial standard for HIC chromatography during recent decades.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty