Would you like to log in again?
ANX Sepharose™ 4 Fast Flow (High Sub) Ion Exchange Chromatography Media, Cytiva
Supplier: Cytiva
Sepharose™
17128710 17-1287-10 17128701 17-1287-01
97067-984EA 1006.45 USD InStock
97067-984 97067-988 CA97067-984 CA97067-988
ANX Sepharose™ 4 Fast Flow (High Sub) Ion Exchange Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
ANX Sepharose™ 4 Fast Flow (high sub) is a weak anion exchanger, with a different selectivity compared with traditional ion exchangers.
  • Weak anion exchanger with different selectivity compared with traditional ion exchangers
  • Applicable for separation of high molecular weight proteins
  • Developed in co-operation with leading large-scale pharmaceutical manufacturers
  • High chemical stability allows for well proven CIP and sanitization protocols
  • The hydrophilic nature of the base matrix ensures low levels of non-specific binding leading to low levels of host cell-derived impurities in the elution pool
  • Stable in all commonly used aqueous buffers
The diethylaminopropylgroup is coupled in a mode that precludes formation of quaternary groups, which are typically seen on traditional DEAE media, resulting in a truly weak anion exchanger. The base matrix, Sepharose 4 Fast Flow, is a highly cross-linked, 4% agarose derivative with high chemical and physical stability and broad separation range. ANX Sepharose 4 Fast Flow (high sub) has high chemical stability, allowing well proven cleaning-in-place (CIP) and sanitization protocols. As member of the BioProcess media range, ANX Sepharose 4 Fast Flow (high sub) meets industrial demands with security of supply and comprehensive technical and regulatory support.
It is optimized for the separation of larger proteins. ANX Sepharose Fast Flow (high sub) is part of the Sepharose Fast Flow ion exchange platform, which has been the industrial standard for ion exchange chromatography during recent decades. ANX Sepharose 4 Fast Flow (high sub) is a weak anion exchanger medium with tertiary amine groups attached to the base matrix Sepharose 4 Fast Flow via ether linkage and a hydrophilic spacer arm
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty