Would you like to log in again?
HiTrap Columns, Capto IEX Selection Kit, Cytiva
Supplier: Cytiva
HiTrap™
28-9343-88 28934388
97068-042EA 455.09 USD InStock
97068-042 CA97068-042
HiTrap Columns, Capto IEX Selection Kit, Cytiva
Chromatography Columns
HiTrap Capto IEX Selection Kit is designed for screening the optimal Capto ion exchange or Cytiva multimodal chromatography resin prior to scale-up.
  • Fast and easy selection. Five prepacked HiTrap columns, each containing a different IEX resin, including multimodal ion exchangers
  • Convenient way to screen for the optimal Cytiva IEX resin for a given application
  • Excellent for optimization studies, buffer composition, pH, flow rate, and other parameters can be optimized with small samples before scale-up
  • HiTrap column format is designed for use with a syringe, peristaltic pump, or chromatography system such as an ÄKTA system
This kit contains a variety of columns prepacked with anion and cation exchange resins, allowing users to screen for the most appropriate column prior to scale-up.
Delivery information: Kit includes five 1 ml HiTrap™ columns prepacked with Capto™ Q (strong anion), Capto™ S (strong cation), Capto™ DEAE (weak anion), Capto™ MMC (multimodal cation), Capto™ adhere (multimodal strong anion), as well as connectors and instructions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty