Would you like to log in again?
HiTrap Columns, Con A 4B, Cytiva
Supplier: Cytiva
HiTrap™
28952085 28-9520-96 28-9520-85 28952096
97068-046PK 327.49 USD InStock
97068-046 CA97068-046 97068-048 CA97068-048
HiTrap Columns, Con A 4B, Cytiva
Chromatography Columns
HiTrap™ Con A 4B is a ready-to-use column prepacked with Con A Sepharose™ 4B, a medium for convenient separation and purification of glycoproteins, polysaccharides, and glycolipids.
  • Con A Sepharose™ is concanavalin A coupled to Sepharose™ 4B
  • Can be operated with a syringe, a peristaltic pump, or a liquid chromatography system such as ÄKTA design
Concanavalin A (Con A) is a tetrameric metalloprotein isolated from Canavalia ensiformis (jack bean). Con A binds molecules containing α-D-mannopyranosyl, α-D-glucopyranosyl, and sterically related residues. The binding sugar requires the presence of C-3, C-4 and C-5 hydroxyl groups for reaction with Con A.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty