Would you like to log in again?
Perfringens Agar Base (T.S.C./S.F.P. Agar Base) Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M837-5KG M837-500G M837-2.5KG M837-500G
95022-336EA 291.06 USD InStock
CA95022-336 95022-336 97068-778 97068-780
Perfringens Agar Base (T.S.C./S.F.P. Agar Base) Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the presumptive identification and enumeration of Clostridium perfringens.
  • Microbiological culture media
Requires added selective supplement and enrichment.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty