Would you like to log in again?
Cole-Parmer® RS-200 Series Reaction Stations, Antylia Scientific
Supplier: Antylia Scientific
36630-02 36630-10
97109-362EA 10073.15 USD InStock
97109-362 97109-356
Cole-Parmer® RS-200 Series Reaction Stations, Antylia Scientific
Stirrers Magnetic Hotplate Stirrers
Run up to 10 reactions simultaneously.
  • Modular design accommodates a variety of cartridges and tubes
  • Control and maintain the same temperature and stirring rate at each position
  • Set stirring speeds from 100 to 800 rpm
  • Simple menus and touch pad interface make controlling quick and easy
  • Compact footprint allows use under fume hood
  • Exterior remains cool to the touch for added safety
Conduct parallel synthesis for 10 reactions simultaneously at the same controlled temperature and stirring rate. All heating blocks, glassware, and reflux heads are interchangeable to accommodate sample volumes ranging from 1 to 1000 ml.
All heater cartridges feature 'in block heaters' for temperature up to 220 °C. Use with the included temperature probe for ±0.9 °F (±0.5 °C) temperature accuracy. Connect cooling plug 36630-71 and chiller to the cartridge to achieve temperature down to –22 °F (–30 °C) without the need for an ice bath. A central gas port, combined with gas-tight PTFE caps, allows a vacuum to be pulled or an inert gas to be introduced.
Delivery information: Supplied with reflux inerting head (36630-72), 10 glass vessels (36630-85), 10 inerting caps (36630-74), 10 stir bars (36630-98), temperature probe (36630-70), one pack of cut tubing, and a power cord with US plug.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty