Would you like to log in again?
Three-Shelf Stainless Steel Utility Carts, Bandy
Supplier: Bandy
CRT04-04 CRT04-07 CRT04-08 CRT04-09 CRT04-02 CRT04-06 CRT04-05 CRT04-03 CRT04-01
98000-568EA 2890.83 USD InStock
98000-568 98000-578 98000-566 98000-576 98000-570 98000-564 98000-574 98000-562 98000-572
Three-Shelf Stainless Steel Utility Carts, Bandy
Carts
Constructed of Type 304 stainless steel with a No
4 satin finish. Carts feature a perforated top with 9.5mm (3/8") holes, 2.5cm (1") round tube handle, four swivel casters (two with locking brakes), and a 2.5cm (1") high fence on three sides. Height: 99.1cm (39").
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty