Would you like to log in again?
Cleanroom Dispenser with One Compartment, S-Curve Technologies
Supplier: S-Curve Technologies
AT-100 PETG AT-100 C
98106-902CS 472.42 USD InStock
98106-902 89108-030
Cleanroom Dispenser with One Compartment, S-Curve Technologies
PPE Dispensers
Designed to protect gloves, bouffants, shoe covers, etc
Easy access and assures first in/first out usage.
Wall mounts or free-standing/wall mount bracket on back.
Flame polished edges for smoothness.
Unique dispensing port provides.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty