Would you like to log in again?
DISPENSER WIPER PETG
Supplier: S-CURVE TECHNOLOGIES
Wiper Dispenser^ 1/4 in clear petg material-safe with ipa. designed for 9 x 9 wipes. no lid-easy access. mounts to wall or free-standing. 1 per case
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
DISPENSER WIPER PETG 98106-936 WDNL-99 PETG Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty