Would you like to log in again?
PCR and qPCR Consumables, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Agilent
600567 410098 410186 401334 410091 410152 410152 410094 401333 410187 410187 410088 410188 410094 410092 410186 401334 401411 410098 410188 410090 401333 410092 600567 410091 401411 410088 410090
76204-934PK 137.1 USD InStock
76204-934 76204-936 76204-938 97066-460 99901-868 CA99901-854 CA99901-852 CA99901-856 ST401333 CA99900-140 99900-152 CAST401333 CA76204-934 CAST401334 ST401334 CA97066-460 ST410094 CA76204-938 99901-860 CA76204-936 99901-852 CA99901-860 99901-856 99901-854 CA99900-152 CA99901-868 99900-140 CAST410094
PCR and qPCR Consumables, Agilent Technologies
Thermal Cyclers
High-quality well plate supplies and accessories will enhance your PCR and qPCR workflow and analysis.
Agilent AriaMx low profile strip tubes are optimized for PCR and qPCR applications. The tubes are validated for optimal performance with Mx3005P, Mx3000P, and the AriaMx real time PCR Systems. Part of the qPCR Plastics and Accessories, the products include the perfect fit frame, tubes, and caps.
Additional products available include the 96-well semiskirted polypropylene PCR plate compatible with the optical Strip Caps, the StrataCoolers LP Benchtop Cooler, and StrataCooler cryo preservation module. Tubes and accessories are compatible most PCR and Real-Time instruments.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty