Would you like to log in again?
Tissue Bond, Diagnostic Biosystems
Supplier: Diagnostic BioSystems
TissueBond™
K013 K013
99990-076EA 143.2 USD InStock
99990-076 CA99990-076
Tissue Bond, Diagnostic Biosystems
Stains Microscopy and staining reagents
Proper tissue adhesion to the glass slides is critical in the immunostaining process. Improper adhesion of tissue sections can lead to poor quality staining or even loss of tissue sections during the process of immunostaining. White glue, albumen, chromegelatin and poly-L-lysine are used as tissue adhesives.
Glass treated with TissueBond™ can be used for both paraffin as well as frozen tissue sections. Since epitope recovery may require harsh treatment with proteolytic enzymes, microwave treatment or treatment with saponin etc., there is a need for a tissue adhesive which can irreversibly bind tissue sections to the glass without creating any background.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty