Would you like to log in again?
Fluoromount™ Mounting Media, Diagnostic Biosystems
Supplier: Diagnostic BioSystems
Fluoromount™
K024-200 K024 K024 K024-200
99990-086EA 105.09 USD InStock
99990-086 CA99990-086 CA10098-626 10098-626
Fluoromount™ Mounting Media, Diagnostic Biosystems
Stains Microscopy and staining reagents
Fluoromount is an innovative, aqueous-based mounting medium designed especially for the permanent preservation of fluorescent specimens.
Fluoromount is to be used with tissue sections stained with a multitude of fluorescent dyes. This is compatible with FITC, phycoerythrin, phycocyanin and allophycocyanin. It can also be used with other fluorescent markers such as texas red and rhodamine.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty