Would you like to log in again?
Proteinase K
Supplier: Diagnostic BioSystems
K030 K030
99990-090EA 151.53 USD InStock
99990-090 CA99990-090
Proteinase K
Enzymes
Proteinase K is specifically formulated to achieve proteolytic digestion of formalin fixed, paraffin embedded tissue sections prior to performing in situ hybridization procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty