Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trioctylamine 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Tri-n-octylamine

A15067-AE A15067-AP A15067-0F A15067-0F
AAA-A15067-0FEA 969.99 USD InStock
AAA-A15067-0F AA-A15067-AP AA-A15067-AE CAAA-A15067-0F
Trioctylamine 95%
Trioctylamine


Formula: C₂₄H₅₁N
MW: 353.68 g/mol
Boiling Pt: 164…168 °C/0.7 mm
Melting Pt: –39 °C
Density: 0.810
Flash Pt: 158 °C(316 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00009560
CAS Number: 1116-76-3
EINECS: 214-242-1
UN: 3082
ADR: "9",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty