Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zirconium ≥98.5%, Powder -325 mesh

Supplier: Thermo Scientific Chemicals
Danger

000418-22 000418-14 000418-88 000418-36
AA00418-22EA 316.99 USD InStock
AA00418-22 AA00418-88 AA00418-14 AA00418-36
Zirconium ≥98.5%, Powder -325 mesh
Zirconium


Formula: Zr
MW: 91.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9
UN: 1358
ADR: 4.1,II
Merck Index: 12,10306
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty