Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cesium fluoride ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

010019-88 10019-14 010019-14 010019-22 10019-22 10019-88
AA10019-88EA 27.39 USD InStock
AA10019-88 AA10019-22 AA10019-14 CAAA10019-14 CAAA10019-22 CAAA10019-88
Cesium fluoride ≥99.9% (metals basis)
Cesium fluoride


Formula: CsF
MW: 151.9 g/mol
Boiling Pt: 1251 °C
Melting Pt: 682 °C
Density: 4,627 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010960
CAS Number: 13400-13-0
EINECS: 236-487-3
UN: 2923
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty