Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium 99+% (metals basis), rods (max. 0.8% Sr), Diameter 22 mm (0.9 in)

Supplier: Thermo Scientific Chemicals
Danger

Packaged under argon.

010103-36 10103-36 10103-22 010103-22 10103-A1 010103-A1
AA10103-22EA 198.77 USD InStock
CAAA10103-22 CAAA10103-A1 AA10103-22 AA10103-A1 CAAA10103-36 AA10103-36
Barium 99+% (metals basis), rods (max. 0.8% Sr), Diameter 22 mm (0.9 in)
Barium
Used as getter, in cathode ray tubes, thermionic emitter tubes and sodium vapor lamps


Formula: Ba
Boiling Pt: 1640 °C (1013 hPa)
Melting Pt: 725 °C
Density: 3.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134031
CAS Number: 7440-39-3
EINECS: 231-149-1
UN: 1400
ADR: 4.3,II
Merck Index: 12,00991
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty