Synonyms:
Platinium
10283-MAT
10283-MAT
010283-FF
10283-FI
010283-GH
010283-FI
10283-FF
10283-GH
10283-FL
10283-FL
AA10283-MATEA
1281.61
USD
InStock
CAAA10283-FF
AA10283-MAT
AA10283-FF
CAAA10283-GH
CAAA10283-FI
AA10283-GH
CAAA10283-MAT
CAAA10283-FL
AA10283-FI
AA10283-FL
Platinum ≥99.9% (metals basis), gauze woven from wire 52 mesh, Diameter 0.1 mm (0.004 in)
Platinum
Platinum gauze, 52 mesh woven from 0.1 mm (0.004") dia. wire.