Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diammonium hydrogen orthophosphate P₂O₅ ≥ 53%, tech.

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: di-Ammonium hydrogen phosphate Diammonium hydrogen orthophosphate Ammonium monohydrogen phosphate Ammonium phosphate dibasic Ammonium hydrogen phosphate

10301-A1 010301-A7 010301-A1 10301-A7
AA10301-A7EA 187.99 USD InStock
CAAA10301-A7 AA10301-A7 AA10301-A1 CAAA10301-A1
Diammonium hydrogen orthophosphate P₂O₅ ≥ 53%, tech.
di-Ammonium hydrogen orthophosphate
Corrosion inhibitors, in fermentation, as flux for soldering tin, copper, brass, and zinc


Formula: (NH₄)₂HPO₄
MW: 132.06 g/mol
Melting Pt: 155 °C (dec.)
Density: 1,619
Storage Temperature: Ambient
MDL Number: MFCD00010891
CAS Number: 7783-28-0
EINECS: 231-987-8
REACH: 01-2119490974-22
Merck Index: 13,00546
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty