Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis(3-aminopropyl)phenylphosphine

Supplier: Thermo Scientific Chemicals
Danger

10427.09 10427-09
AA10427-09EA 414.99 USD InStock
CAAA10427-09 AA10427-09
Bis(3-aminopropyl)phenylphosphine
Bis(3-aminopropyl)phenylphosphine


Formula: (H₂NC₃H₆)₂P(C₆H₅)
MW: 224.29 g/mol
Boiling Pt: 144 °C (1 mmHg)
Density: 1.03
Storage Temperature: Ambient
MDL Number: MFCD00014834
CAS Number: 6775-01-5
UN: 3267
ADR: 8,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty