Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium oxide, γ-α-phase ≥99.98% (metals basis)

Supplier: Thermo Scientific Chemicals

10459-14 10459-22 010459-14 010459-36 10459-36 010459-22
AA10459-22EA 123.99 USD InStock
AA10459-22 CAAA10459-22 AA10459-14 AA10459-36 CAAA10459-14 CAAA10459-36
Aluminium oxide, γ-α-phase ≥99.98% (metals basis)
Aluminium oxide, γ-α-phase


Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980 °C
Melting Pt: 2045 °C
Density: 3.965
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
Merck Index: 12,00369
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty