Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

cis-Platin Pt ≥ 64.5% for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: cis-Diamminedichloroplatinum (II) cis-Diammineplatinum (II) dichloride Platinol

010471-01 010471-03 10471-06 010471-06 10471-01 10471-03
AA10471-06EA 1964.99 USD InStock
AA10471-06 AA10471-03 CAAA10471-06 CAAA10471-01 AA10471-01 CAAA10471-03
cis-Platin Pt ≥ 64.5% for NMR spectroscopy
cis-Platin
Potent anticancer agent that blocks DNA synthesis


Formula: H₆Cl₂N₂Pt
MW: 300.05 g/mol
Melting Pt: 270 °C (dec.)
Density: 3.738 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011623
CAS Number: 15663-27-1
EINECS: 239-733-8
UN: 3288
ADR: 6.1,II
Merck Index: 13,02341
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty