10484.06
10484-06
10484.03
10484-03
AA10484-06EA
481.99
USD
InStock
CAAA10484-06
AA10484-06
AA10484-03
CAAA10484-03
trans-Dichlorobis(diethylsulfide)platinum(II) (43,7% Pt)
trans-Dichlorobis(diethylsulfide)platinum(II)
Applications: Hydrosilation
Soluble in acetone, alcohol and benzene