Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dodecacarbonyltriruthenium 99%

Supplier: Thermo Scientific Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Ruthenium carbonylTriruthenium dodecacarbonyl

10507-03 010507-03 010507-06 10507-06
AA10507-03EA 128.99 USD InStock
CAAA10507-03 AA10507-03 CAAA10507-06 AA10507-06
Dodecacarbonyltriruthenium 99%
Dodecacarbonyltriruthenium
Application: Carbonylation

Sparingly soluble in hydrocarbons (e.g. hexane, cyclohexane, benzene) and acetone. Solutions undergo some decomposition on strong heating


Formula: C₁₂O₁₂Ru₃
MW: 639.33 g/mol
Melting Pt: ∼ 150 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011209
CAS Number: 15243-33-1
EINECS: 239-287-4
UN: 3466
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty