Synonyms:
Ruthenium carbonyl
, Triruthenium dodecacarbonyl
10507-03
010507-03
010507-06
10507-06
AA10507-03EA
128.99
USD
InStock
CAAA10507-03
AA10507-03
CAAA10507-06
AA10507-06
Dodecacarbonyltriruthenium 99%
Dodecacarbonyltriruthenium
Application: Carbonylation
Sparingly soluble in hydrocarbons (e.g. hexane, cyclohexane, benzene) and acetone. Solutions undergo some decomposition on strong heating