010536-03
10536-03
10536-06
010536-06
AA10536-03EA
172.99
USD
InStock
AA10536-03
CAAA10536-03
AA10536-06
CAAA10536-06
Potassium tetracyanoplatinate(II) trihydrate (≥44,9% Pt) ≥99.9% (metals basis)
Potassium tetracyanoplatinate (II) trihydrate
Potassium tetracyanoplatinate(II) trihydrate is used in the preparation other salts like Krogmann's salt, that are highly conducting, one dimensional materials.
Soluble in hot water