Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminum phosphate ≥99.99% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Aluminium phosphate

010628-06 010628-14 10628-06 10628-14
AA10628-14EA 366.99 USD InStock
AA10628-14 CAAA10628-14 AA10628-06 CAAA10628-06
Aluminum phosphate ≥99.99% (metals basis), Puratronic®
Aluminum phosphate


Formula: AlPO₄
MW: 121.951 g/mol
Melting Pt: >1500 °C
Density: 2.566
Storage Temperature: Ambient
MDL Number: MFCD00003430
CAS Number: 7784-30-7
EINECS: 232-056-9
UN: 3260
ADR: 8,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty