Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium chloride hydrate ≥99.9965% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

10680-30 010680-09 10680-04 010680-30 10680-18 010680-18 10680-09 010680-04
AA10680-30EA 1654.99 USD InStock
AA10680-30 CAAA10680-30 CAAA10680-18 AA10680-04 CAAA10680-04 AA10680-09 AA10680-18 CAAA10680-09
Calcium chloride hydrate ≥99.9965% (metals basis), Puratronic®
Calcium chloride hydrate


Formula: CaCl₂·6H₂O
MW: 110.98 g/mol (anhy)
Melting Pt: 29.54 °C
Density: 1.71
Storage Temperature: Ambient
MDL Number: MFCD00149616
CAS Number: 7774-34-7
EINECS: 607-129-7
Merck Index: 13,01660
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty