Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) chloride dihydrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Copper dichloride dihydrate Cupric chloride dihydrate

010698-14 10698-06 010698-06 10698-14 010698-22 10698-22
AA10698-06EA 46.87 USD InStock
AA10698-06 CAAA10698-14 CAAA10698-06 AA10698-14 CAAA10698-22 AA10698-22
Copper(II) chloride dihydrate ≥99.999% (metals basis), Puratronic®
Copper (II) chloride dihydrate


Formula: CuCl₂·2H₂O
MW: 170.48 g/mol
Melting Pt: 100 °C (dec.)
Density: 2.510
Storage Temperature: Ambient
MDL Number: MFCD00149674
CAS Number: 10125-13-0
EINECS: 231-210-2
UN: 2802
ADR: 8,III
Merck Index: 13,02660
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty