Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium iodide hydrate ≥99.995% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Lithium iodide hydrate

10738-14 10738-06 010738-14 010738-06
AA10738-06EA 64.59 USD InStock
CAAA10738-06 AA10738-06 CAAA10738-14 AA10738-14
Lithium iodide hydrate ≥99.995% (metals basis), Puratronic®
Lithium iodide hydrate
Lithium iodide hydrate is used as a reagent for ester cleavage and decarboxylation and C-C bond forming reactions. It is also used as source of nucleophilic iodide, as a mild lewis acid, epoxide opening and as an additive for organometallic-mediated transformations.


Formula: ILi·H₂O
MW: 133.84 g/mol (anhy)
Melting Pt: 73 °C (-H₂O)
Storage Temperature: Ambient
MDL Number: MFCD00149769
CAS Number: 17023-25-5
EINECS: 233-822-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty