Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium acetate tetrahydrate ≥99.997% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Synonyms: Acetic acid magnesium salt tetrahydrate Magnesium di(acetate) tetrahydrate

010795-14 10795-14 010795-22 10795-22
AA10795-22EA 563.33 USD InStock
CAAA10795-22 AA10795-22 CAAA10795-14 AA10795-14
Magnesium acetate tetrahydrate ≥99.997% (metals basis), Puratronic®
Magnesium acetate tetrahydrate


Formula: (H₃CCOO)₂Mg·4H₂O
MW: 214.46 g/mol
Melting Pt: 80 °C
Density: 1.454
Storage Temperature: Ambient
MDL Number: MFCD00149214
CAS Number: 16674-78-5
EINECS: 205-554-9
Merck Index: 13,05676
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty