Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium hydroxide carbonate hydrate ≥99.996% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Synonyms: Magnesium carbonate hydroxide Magnesium carbonate hydrated basic Magnesium carbonate basic

10796-06 010796-22 10796-22 010796-14 010796-06 10796-14
AA10796-22EA 919.99 USD InStock
CAAA10796-22 CAAA10796-14 AA10796-22 AA10796-14 CAAA10796-06 AA10796-06
Magnesium hydroxide carbonate hydrate ≥99.996% (metals basis), Puratronic®
Magnesium hydroxide carbonate hydrate


Formula: MgCO₃·Mg(OH)₂·xH₂O
MW: 142.63 g/mol (anhy)
Boiling Pt: 3600 °C (1013 hPa)
Melting Pt: Dec.
Density: 2.16 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149783
CAS Number: 12125-28-9
EINECS: 235-192-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty