Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Manganese(II) acetate tetrahydrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Acetic acid manganese (II) salt tetrahydrate Manganese diacetate tetrahydrate

10802-14 010802-06 10802-06 010802-14
AA10802-14EA 194.99 USD InStock
AA10802-14 CAAA10802-06 CAAA10802-14 AA10802-06
Manganese(II) acetate tetrahydrate ≥99.999% (metals basis), Puratronic®
Manganese (II) acetate tetrahydrate


Formula: (H₃CCOO)₂Mn·4H₂O
MW: 245.09 g/mol
Melting Pt: 80 °C
Density: 1.589
Storage Temperature: Ambient
MDL Number: MFCD00062552
CAS Number: 6156-78-1
EINECS: 211-334-3
Merck Index: 13,05746
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty