Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Manganese(II) nitrate hydrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Manganese dinitrate hydrate Manganous nitrate hydrate

10806-04 10806-09 010806-09 010806-04 010806-18 10806-18
AA10806-09EA 77.59 USD InStock
AA10806-09 CAAA10806-09 AA10806-18 CAAA10806-18 AA10806-04 CAAA10806-04
Manganese(II) nitrate hydrate ≥99.999% (metals basis), Puratronic®
Manganese (II) nitrate hydrate


Formula: Mn(NO₃)₂·H₂O
MW: 196.96 g/mol
Melting Pt: ∼ 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00149788
CAS Number: 15710-66-4
EINECS: 233-828-8
UN: 2724
ADR: 5.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty