Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mercury(II) chloride ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Mercuric chloride Mercury dichloride

10808-14 010808-22 10808-22
AA10808-22EA 377 USD InStock
CAAA10808-22 AA10808-22 CAAA10808-14
Mercury(II) chloride ≥99.999% (metals basis), Puratronic®
Mercury (II) chloride


Formula: HgCl₂
MW: 271.5 g/mol
Boiling Pt: 302 °C
Melting Pt: 280…282 °C
Density: 5.4
Storage Temperature: Ambient
MDL Number: MFCD00011041
CAS Number: 7487-94-7
EINECS: 231-299-8
UN: 1624
ADR: 6.1,II
Merck Index: 13,05901
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty