Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium molybdate (para) hydrate ≥99.999% (metals basis excluding W), Puratronic® (max. 300 ppm W)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Hexaammonium heptamolybdate Ammonium paramolybdate Ammonium molybdate (para)

10811.06 10811-14 10811.14 10811-06
AA10811-14EA 247 USD InStock
AA10811-14 CAAA10811-14 AA10811-06 CAAA10811-06
Ammonium molybdate (para) hydrate ≥99.999% (metals basis excluding W), Puratronic® (max. 300 ppm W)
Ammonium molybdate (para) hydrate


Formula: H₂₄Mo₇N₆O₂₄
MW: 1163.8 g/mol (anhy)
Melting Pt: ∼ 190 °C (dec.)
Density: 2.498
Storage Temperature: Ambient
MDL Number: MFCD00064633
CAS Number: 12027-67-7
EINECS: 234-722-4
Merck Index: 13,00536
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty