Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nickel acetate tetrahydrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Nickel di(acetate) tetrahydrate Acetic acid nickel (II) salt tetrahydrate Nickel (II) acetate tetrahydrate Nickel diacetate tetrahydrate

010813-14 10813-06 010813-06 10813-14
AA10813-14EA 469.67 USD InStock
AA10813-14 CAAA10813-06 CAAA10813-14 AA10813-06
Nickel acetate tetrahydrate ≥99.999% (metals basis), Puratronic®
Nickel acetate tetrahydrate


Formula: C₄H₆NiO₄·4H₂O
MW: 248.84 g/mol
Boiling Pt: 16 °C (1013 hPa)
Melting Pt: Dec.
Density: 1.744
Storage Temperature: Ambient
MDL Number: MFCD00066973
CAS Number: 6018-89-9
EINECS: 206-761-7
UN: 3077
ADR: 9,III
Merck Index: 12,06583
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty