Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium chloride ≥99.997% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Potassiumchloride

010839-30 010839-18 10839-30 10839-18
AA10839-18EA 145.99 USD InStock
CAAA10839-18 AA10839-18 AA10839-30 CAAA10839-30
Potassium chloride ≥99.997% (metals basis), Puratronic®
Potassium chloride
Powder. Hygroscopic. Purity based on metal contaminates only.


Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1500 °C (subl.)
Melting Pt: 773 °C
Density: 1.984
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 13,07704
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty