Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Selenious acid ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Selenous acid

010855-14 010855-22 10855-14 10855-22
AA10855-22EA 1083.99 USD InStock
CAAA10855-14 AA10855-22 AA10855-14 CAAA10855-22
Selenious acid ≥99.999% (metals basis), Puratronic®
Selenious acid


Formula: H₂O₃Se
MW: 128.97 g/mol
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 70 °C (dec.)
Density: 1.998 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011331
CAS Number: 7783-00-8
EINECS: 231-974-7
UN: 3290
ADR: 6.1,II
Merck Index: 14,08430
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty